O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98+%
O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98+%
O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98+%
O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98+%
Thermo Scientific Chemicals

O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98+%

CAS: 330645-87-9 | C11H15ClF6N5OP | 413.689 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number H26402.09
also known as H26402-09
Price (USD)
55.65
Online Exclusive
61.60
Save 5.95 (10%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.60
Save 5.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS330645-87-9
IUPAC Namehexafluoro-λ⁵-phosphanuide; {[(6-chloro-1H-1,2,3-benzotriazol-1-yl)oxy](dimethylamino)methylidene}dimethylazanium
Molecular FormulaC11H15ClF6N5OP
InChI KeyZHHGTMQHUWDEJF-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CN(C)C(ON1N=NC2=CC=C(Cl)C=C12)=[N+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤0.5%
FormPowder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (HPLC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only