1-Bromo-2-ethylbutane, 97%
1-Bromo-2-ethylbutane, 97%
1-Bromo-2-ethylbutane, 97%
Thermo Scientific Chemicals

1-Bromo-2-ethylbutane, 97%

CAS: 3814-34-4 | C6H13Br | 165.074 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H26445.AA
also known as H26445-AA
Price (USD)
25.30
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
25.30
Each
Chemical Identifiers
CAS3814-34-4
IUPAC Name3-(bromomethyl)pentane
Molecular FormulaC6H13Br
InChI KeyKKGUMGWNFARLSL-UHFFFAOYSA-N
SMILESCCC(CC)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4490-1.4540 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only