6-Methoxypyridine-3-carboxaldehyde, 98%
6-Methoxypyridine-3-carboxaldehyde, 98%
6-Methoxypyridine-3-carboxaldehyde, 98%
Thermo Scientific Chemicals

6-Methoxypyridine-3-carboxaldehyde, 98%

CAS: 65873-72-5 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26515.06
also known as H26515-06
Price (USD)
213.65
Special offer
Online exclusive
Ends: 30-Jun-2026
251.00
Save 37.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
213.65
Special offer
Online exclusive
Ends: 30-Jun-2026
251.00
Save 37.35 (15%)
Each
Chemical Identifiers
CAS65873-72-5
IUPAC Name6-methoxypyridine-3-carbaldehyde
Molecular FormulaC7H7NO2
InChI KeyCTAIEPPAOULMFY-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)47.0-53.0?C
Appearance (Color)Dark cream to cream to pale brown
Free acid (titration)≤1%
Assay (GC)≥97.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only