1-Benzyl-1,4,7,10-tetraazacyclododecane
1-Benzyl-1,4,7,10-tetraazacyclododecane
1-Benzyl-1,4,7,10-tetraazacyclododecane
Thermo Scientific Chemicals

1-Benzyl-1,4,7,10-tetraazacyclododecane

CAS: 112193-83-6 | C15H26N4 | 262.401 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H26525.03
also known as H26525-03
Price (USD)
422.65
Online Exclusive
469.00
Save 46.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
422.65
Online Exclusive
469.00
Save 46.35 (10%)
Each
Chemical Identifiers
CAS112193-83-6
IUPAC Name1-benzyl-1,4,7,10-tetraazacyclododecane
Molecular FormulaC15H26N4
InChI KeyFURLCQRFFWBENR-UHFFFAOYSA-N
SMILESC(N1CCNCCNCCNCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (HPLC)≥95.0%
Proton NMRConforms to structure
Used as ligands for catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as ligands for catalysts.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. F Chuburu.; M Le Baccon.; H Handel. Stoichiometric mono N-functionalization of cyclen via a boron protected intermediate. Tetrahedron. 2001, 57 (12), 2385-2390.
  2. Stephan W. Kohl.; Katharina Kuse.; Markus Hummert.; Herbert Schumann.; Clemens Mügge.; Katharina Janek.; Hardy Weißhoff. New Synthetic Routes for 1-Benzyl-1,4,7,10-tetraazacyclododecane and 1,4,7,10-Tetraazacyclododecane-1-acetic Acid Ethyl Ester, Important Starting Materials for Metal-coded DOTA-Based Affinity Tags. Zeitschrift für Naturforschung B. 2007, 62 (3), 397-406.