2-Aminoisobutyric acid methyl ester hydrochloride, 99%
2-Aminoisobutyric acid methyl ester hydrochloride, 99%
2-Aminoisobutyric acid methyl ester hydrochloride, 99%
2-Aminoisobutyric acid methyl ester hydrochloride, 99%
Thermo Scientific Chemicals

2-Aminoisobutyric acid methyl ester hydrochloride, 99%

CAS: 15028-41-8 | C5H12ClNO2 | 153.606 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26550.03
also known as H26550-03
Price (USD)
61.65
Online Exclusive
68.80
Save 7.15 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
61.65
Online Exclusive
68.80
Save 7.15 (10%)
Each
Add to cart
Chemical Identifiers
CAS15028-41-8
IUPAC Namemethyl 2-amino-2-methylpropanoate hydrochloride
Molecular FormulaC5H12ClNO2
InChI KeyNVWZNEDLYYLQJC-UHFFFAOYSA-N
SMILESCl.COC(=O)C(C)(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
Methyl α-aminoisobutyrate is used as a specific substrate of amino acid transport system and in studies on amino acid transport systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl α-aminoisobutyrate is used as a specific substrate of amino acid transport system and in studies on amino acid transport systems.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Martina Wurdemann.; Jens Christoffers. Nitrile Oxide versus Nitrone - Switching Regioselectivity of Intramolecular 1,3-Dipolar Cycloadditions towards the Preparation of Aminopiperidinecarboxylates. European Journal of Organic Chemistry. 2013, 2013, (32), 7421-7431.