Casein Peptone
Thermo Scientific Chemicals

Casein Peptone

Casein peptone is an enzymatic derivative of casein, CAS # 9000-71-9, the protein found in milk and animal tissues.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number H26557.36
also known as H26557-36
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS91079-40-2
SpecificationsSpecification SheetSpecification Sheet
pH7.0 ? 0.5 (2% solution in water)
Appearance (Color)White to cream or pale brown
FormPowder
Amino Nitrogen≥4%
Total Nitrogen≥10%
Casein peptone is used in nutrient media for growing bacteria and fungi. It is employed in antibiotics and enzyme productions. It is also essential for analytical microbiology and industrial fermentation process. It is also used in bacterial diagnostics and in agro-food, cosmetic and pharmaceutical fermentation for production of biomass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Casein peptone is commonly used in the preparation of culture media in microbiology

• Casein peptone is an enzymatically prepared hydrolysate of casein

• Casein peptone is an opioid peptide and a rich source of amino acids

Application

• Supports the growth of a plethora of organisms in culture, particularly in serum-free media

• Enzymatic digest of casein peptone can provide active peptides for research on opioid signaling

RUO – Research Use Only

General References:

  1. Covas C, Almeida B, Esteves AC, Lourenço J, Domingues P, Caetano T, Mendo S. Peptone from casein, an antagonist of nonribosomal peptide synthesis: a case study of pedopeptins produced by Pedobacter lusitanus NL19. N Biotechnol. 2021 Jan 25;60:62-71. doi: 10.1016/j.nbt.2020.07.006. Epub 2020 Sep 3. PMID: 32891869.
  2. Davami F, Eghbalpour F, Nematollahi L, Barkhordari F, Mahboudi F. Effects of Peptone Supplementation in Different Culture Media on Growth, Metabolic Pathway and Productivity of CHO DG44 Cells; a New Insight into Amino Acid Profiles. Iran Biomed J. 2015;19(4):194-205. doi: 10.7508/ibj.2015.04.002. Epub 2015 Aug 1. PMID: 26232332; PMCID: PMC4649854.