N,N'-Disuccinimidyl carbonate, tech. 85%, remainder N-Hydroxysuccinimide
N,N'-Disuccinimidyl carbonate, tech. 85%, remainder N-Hydroxysuccinimide
N,N'-Disuccinimidyl carbonate, tech. 85%, remainder N-Hydroxysuccinimide
Thermo Scientific Chemicals

N,N'-Disuccinimidyl carbonate, tech. 85%, remainder N-Hydroxysuccinimide

CAS: 74124-79-1 | C9H8N2O7 | 256.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H26629.14
also known as H26629-14
Price (USD)
331.65
Online Exclusive
368.00
Save 36.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
331.65
Online Exclusive
368.00
Save 36.35 (10%)
Each
Chemical Identifiers
CAS74124-79-1
IUPAC Namebis(2,5-dioxopyrrolidin-1-yl) carbonate
Molecular FormulaC9H8N2O7
InChI KeyPFYXSUNOLOJMDX-UHFFFAOYSA-N
SMILESO=C(ON1C(=O)CCC1=O)ON1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (HPLC)(N-Hydroxysuccinimide derivatives) : ≥99.0%
Identification (FTIR)Conforms
N,N'-Disuccinimidyl carbonate is used to prepare cleavable linker succinimide vinylsufonylethyl carbonate for thiol modified-DNA. It serves as a reagent for the synthesis of carbamates. Further, it is used as a coupling reagent as well as in the preparation of N-succinimidyl esters of N-protected amino acids, activated carbonate and coupling of ligand to proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Disuccinimidyl carbonate is used to prepare cleavable linker succinimide vinylsufonylethyl carbonate for thiol modified-DNA. It serves as a reagent for the synthesis of carbamates. Further, it is used as a coupling reagent as well as in the preparation of N-succinimidyl esters of N-protected amino acids, activated carbonate and coupling of ligand to proteins.

Solubility
Soluble in dimethyl sulfoxide, hot pyridine, acetonitrile and most organic solvents. Insoluble in water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong acids and oxidizing agents.
RUO – Research Use Only

General References:

  1. Černoch, P.; Černochová, Z.; Kučka, J.; Hrubý, M.; Petrova, S.; Štěpánek, P. Thermoresponsive polymer system based on poly(N-vinylcaprolactam) intended for local radiotherapy applications. Appl. Radiat. Isot. 2015, 98, 7-12.
  2. Nale, D. B.; Bhanage, B. M. Copper-catalyzed efficient synthesis of a 2-benzimidazolone scaffold from 2-nitroaniline and dimethyl carbonate via a hydrosilylation reaction. Green Chem. 2015, 17 (4), 2480-2486.