(+/-)-Propranolol hydrochloride, 99%
(+/-)-Propranolol hydrochloride, 99%
(+/-)-Propranolol hydrochloride, 99%
Thermo Scientific Chemicals

(+/-)-Propranolol hydrochloride, 99%

CAS: 318-98-9 | C16H22ClNO2 | 295.807 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H26645.06
also known as H26645-06
Price (USD)
88.10
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
88.10
Each
Chemical Identifiers
CAS318-98-9
IUPAC Namehydrogen 1-(naphthalen-1-yloxy)-3-[(propan-2-yl)amino]propan-2-ol chloride
Molecular FormulaC16H22ClNO2
InChI KeyZMRUPTIKESYGQW-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(C)NCC(O)COC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (Titration ex Chloride)≥98.5 to ≤101.5% (non-U.S. specification)
Assay≥98.5% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
A beta-adrenergic blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only