How do I prepare 1 L of Thermo Scientific Chemicals Miller LB Broth media (Cat. No. H26676.xx)?
To prepare 1 L of Thermo Scientific Chemicals Miller LB media:
- Add 25 g of powder to 1 L distilled water
- Mix by vortexing
- Sterilize by autoclaving for 25 minutes at 121 degrees C