LB Broth (Miller's modification)
Thermo Scientific Chemicals
LB Broth (Miller's modification)
(+)-Dehydroisoandrosterone, CAS # 53-43-0, also known as androstenolone, is an endogenous steroid hormone precursor. It is an intermediate in the conversion of cholesterol to androgens and estrogens.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging Type:
View all
Plastic Bottle
Catalog number H26676.36
also known as H26676-36
Price (USD)
-
Quantity:
500 g
Packaging Type:
Plastic Bottle
Request bulk or custom format
Specifications
Packaging TypePlastic Bottle
Quantity500 g
Recommended StorageAmbient temperatures
SensitivityHygroscopic
FormPowder
View more
For the growth of E. coli and other enteric bacteria. LB Luria contains less salt than LB Miller and LB Lennox. LB Luria is used for cultivating salt-sensitive strains of E. coli and enteric bacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • (+)-Dehydroisoandrosterone is an endogenous steroid hormone precursor that acts as an intermediate in the conversion of cholesterol to androgens and estrogens
Applications
  • It is an endogenous and naturally occurring steroid hormone produced largely by the adrenal glands, but also by the testes and ovaries
  • It has been described that (+)-Dehydroisoandrosterone could have a role in the immune and stress response
  • (+)-Dehydroisoandrosterone can be converted to testosterone, androstenedione, estradiol, and estrone
  • (+)-Dehydroisoandrosterone is derived from cholesterol through the action of two cytochrome P450 enzymes
  • It can induce both direct and indirect in vivo biological effects, by modulating GABA- α receptor and N-Methyl-D-aspartic acid (NMDA) receptor complexes.
  • It can also increase pancreatic β cell insulin secretion and anti-glucocorticoid activity
  • It is useful to detect excess adrenal activity since it is produced nearly exclusively by the adrenal glands
RUO – Research Use Only
General References:
  1. Prough, R.; Clark, B.; Klinge, C. Novel mechanisms for DHEA action. Journal of Molecular Endocrinology, 2016, 56, 3, R139-R155.
Frequently asked questions (FAQs)
How should I prepare 1 L of Miller LB Broth media (Cat. Nos. 611875000 and H26676.xx)?
To prepare 1 L of Miller LB media:
  1. Add 25 g of powder to 1 L distilled water
  2. Mix by vortexing
  3. Sterilize by autoclaving for 25 minutes at 121 degrees C