2-n-Hexylfuran, 97%
2-n-Hexylfuran, 97%
2-n-Hexylfuran, 97%
Thermo Scientific Chemicals

2-n-Hexylfuran, 97%

CAS: 3777-70-6 | C10H16O | 152.237 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H26698.03
also known as H26698-03
Price (USD)
38.65
Special offer
Online exclusive
Ends: 28-Dec-2026
45.50
Save 6.85
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
38.65
Special offer
Online exclusive
Ends: 28-Dec-2026
45.50
Save 6.85
Each
Chemical Identifiers
CAS3777-70-6
IUPAC Name2-hexylfuran
Molecular FormulaC10H16O
InChI KeyXBLCAKKYMZVLPU-UHFFFAOYSA-N
SMILESCCCCCCC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4480-1.4530 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only