2'-Iodo-1,1':3',1″-terphenyl, 99%
2'-Iodo-1,1':3',1″-terphenyl, 99%
2'-Iodo-1,1':3',1″-terphenyl, 99%
Thermo Scientific Chemicals

2'-Iodo-1,1':3',1″-terphenyl, 99%

CAS: 82777-09-1 | C18H13I | 356.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26704.03
also known as H26704-03
Price (USD)
85.65
Special offer
Online exclusive
Ends: 30-Jun-2026
101.00
Save 15.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
85.65
Special offer
Online exclusive
Ends: 30-Jun-2026
101.00
Save 15.35 (15%)
Each
Chemical Identifiers
CAS82777-09-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)≥98.5%
Melting Point (clear melt)111.0-118.0?C
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only