1,4,7-Tris(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
1,4,7-Tris(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
1,4,7-Tris(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
Thermo Scientific Chemicals

1,4,7-Tris(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane

CAS: 122555-91-3 | C26H50N4O6 | 514.708 g/mol
Have Questions?
Catalog NumberQuantity
H26736.MD
also known as H26736-MD
250 mg
Catalog number H26736.MD
also known as H26736-MD
Price (USD)
178.65
Special offer
Online exclusive
Ends: 30-Jun-2026
210.00
Save 31.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
178.65
Special offer
Online exclusive
Ends: 30-Jun-2026
210.00
Save 31.35 (15%)
Each
Chemical Identifiers
CAS122555-91-3
IUPAC Nametert-butyl 2-{4,7-bis[2-(tert-butoxy)-2-oxoethyl]-1,4,7,10-tetraazacyclododecan-1-yl}acetate
Molecular FormulaC26H50N4O6
InChI KeyNMHVTLJFPDOJOD-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)CN1CCNCCN(CC(=O)OC(C)(C)C)CCN(CC(=O)OC(C)(C)C)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥95.0%
Appearance (Color)White
Proton NMRConforms to structure
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only