2-(Ethoxycarbonyl)ethylzinc bromide, 0.5M in THF
2-(Ethoxycarbonyl)ethylzinc bromide, 0.5M in THF
2-(Ethoxycarbonyl)ethylzinc bromide, 0.5M in THF
2-(Ethoxycarbonyl)ethylzinc bromide, 0.5M in THF
Thermo Scientific Chemicals

2-(Ethoxycarbonyl)ethylzinc bromide, 0.5M in THF

CAS: 193065-68-8 | C5H10BrO2Zn- | 247.417 g/mol
Have Questions?
Catalog NumberQuantity
H26739.AD50 mL
Catalog number H26739.AD
Price (USD)
180.65
Online Exclusive
200.00
Save 19.35 (10%)
Each
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)
180.65
Online Exclusive
200.00
Save 19.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS193065-68-8
IUPAC Namezinc(2+) 3-ethoxy-3-oxopropan-1-ide bromide
Molecular FormulaC5H9BrO2Zn
InChI KeyAPPDFGHVJHZDKS-UHFFFAOYSA-M
SMILES[Zn++].[Br-].CCOC(=O)C[CH2-]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMay form a precipitate on storage
Refractive Index1.4220-1.4300 @20?C
Complexometric Titration0.40M - 0.60M
Appearance (Color)Brown to dark brown
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
In many of the reactions, organozinc compounds appear as intermediates. Some of the synthetic uses of organozinc compounds are: for the stereo selective alkylation reaction of cyclic ketones, conjugate addition reaction to alpha, be
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Knochel, P,; Singer, R. D. “Preparation and Reactions of Polyfunctional Organozinc Reagents in Organic Synthesis. Chem. Rev. 1993, 93 2117-2188 .
  2. Kim, S.-H.; Rieke, R. D. “Recent Advance in Heterocyclic Organozinc and Organomanganese Compounds; Direct Synthetic Routes and Application in Organic Synthesis”.Molecules. 2010, 15 8006-8038..