2-Propionylfuran, 97%
2-Propionylfuran, 97%
2-Propionylfuran, 97%
Thermo Scientific Chemicals

2-Propionylfuran, 97%

CAS: 3194-15-8 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H26754.06
also known as H26754-06
Price (USD)
74.65
Online exclusive
82.80
Save 8.15
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
74.65
Online exclusive
82.80
Save 8.15
Each
Chemical Identifiers
CAS3194-15-8
IUPAC Name1-(furan-2-yl)propan-1-one
Molecular FormulaC7H8O2
InChI KeyHCPORNAVHSWTOJ-UHFFFAOYSA-N
SMILESCCC(=O)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear yellow to brown
Refractive Index1.4975-1.5025 @ 20?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only