4-Cyanobiphenyl, 95%
4-Cyanobiphenyl, 95%
4-Cyanobiphenyl, 95%
Thermo Scientific Chemicals

4-Cyanobiphenyl, 95%

CAS: 2920-38-9 | C13H9N | 179.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number H26791.09
also known as H26791-09
Price (USD)
126.65
Online exclusive
140.00
Save 13.35
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
126.65
Online exclusive
140.00
Save 13.35
Each
Chemical Identifiers
CAS2920-38-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystalline powder or crystals
Assay (GC)≥94.0%
Melting Point83-90?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only