4-Fluoro-2-methoxybenzylamine, 97%
4-Fluoro-2-methoxybenzylamine, 97%
4-Fluoro-2-methoxybenzylamine, 97%
4-Fluoro-2-methoxybenzylamine, 97%
Thermo Scientific Chemicals

4-Fluoro-2-methoxybenzylamine, 97%

CAS: 870563-60-3 | C8H10FNO | 155.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26796.06
also known as H26796-06
Price (USD)
413.65
Online Exclusive
459.00
Save 45.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
413.65
Online Exclusive
459.00
Save 45.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS870563-60-3
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.5220-1.5270 @ 20?C
Appearance (Color)Clear colorless to pale yellow
As pharmaceutical intermediate. As a metabolite of Capsaicin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As pharmaceutical intermediate. As a metabolite of Capsaicin.

Solubility
Insoluble in water.

Notes
Air Sensitive and Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only

General References:

  1. Tamara Meiresonne,; Guido Verniest,; Norbert De Kimpe,; Sven Mangelinckx. Synthesis of 2-Fluoro-1,4-benzoxazines and 2-Fluoro-1,4-benzoxazepin-5-ones by Exploring the Nucleophilic Vinylic Substitution (SNV) Reaction of gem-Difluoroenamides. J. Org. Chem. 2015 , 80 (10) ,5111-5124.
  2. Joseph T. Repinea,; Douglas S. Johnsona,; Timothy Stuka,; Andrew D. Whitea,; Michael A. Stiera,; Tingsheng Lib,; Zhixiang Yangb,; Samarendra N. Maiti. Synthesis of fluorinated 1,8-naphthyridinone derivatives. Tetrahedron Letters. 2007, 48 46,8189-8191.