Agrochemical Intermediates, dyestuff Intermediate, pharmaceutical Intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Agrochemical Intermediates, dyestuff Intermediate, pharmaceutical Intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only
General References:
- A G Zacchei,; L L Weidner,; G H Besselaar,; E B Raftery. Physiological disposition and metabolic fate of a new antiarrhythmic agent, alpha, alpha-dimethyl-4-(alpha, alpha, beta, beta-tetrafluorophenethyl) benzylamine in the rat, dog, monkey, baboon, and man.. DMD. 1976, 4 (4),387-401.
- M L Torchiana,; C A Stone,; H C Wenger,; R Evans,; B Lagerquist,; T O'Malley. The antiarrhythmic and hemodynamic actions of alpha, alpha-dimethyl-4-(alpha, alpha, beta, beta-tetrafluorophenethyl) benzylamine (MK-251). JPET. 1975 , 194 (2), 415-426.