2-Chloro-5-ethylpyrimidine, 98%
2-Chloro-5-ethylpyrimidine, 98%
2-Chloro-5-ethylpyrimidine, 98%
2-Chloro-5-ethylpyrimidine, 98%
Thermo Scientific Chemicals

2-Chloro-5-ethylpyrimidine, 98%

CAS: 111196-81-7 | C6H7ClN2 | 142.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H26856.14
also known as H26856-14
Price (USD)
222.65
Online Exclusive
247.00
Save 24.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
222.65
Online Exclusive
247.00
Save 24.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS111196-81-7
IUPAC Name2-chloro-5-ethylpyrimidine
Molecular FormulaC6H7ClN2
InChI KeyBGLLZQRUXJGTAD-UHFFFAOYSA-N
SMILESCCC1=CN=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow
Assay (GC)≥97.5%
Refractive Index1.5175-1.5215 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only