1-n-Hexyl-4-iodobenzene, 98%
1-n-Hexyl-4-iodobenzene, 98%
1-n-Hexyl-4-iodobenzene, 98%
Thermo Scientific Chemicals

1-n-Hexyl-4-iodobenzene, 98%

CAS: 62150-34-9 | C12H17I | 288.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H26878.03
also known as H26878-03
Price (USD)
38.65
Online Exclusive
42.40
Save 3.75 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
38.65
Online Exclusive
42.40
Save 3.75 (9%)
Each
Chemical Identifiers
CAS62150-34-9
IUPAC Name1-hexyl-4-iodobenzene
Molecular FormulaC12H17I
InChI KeyQCTCAMDTUGSMJX-UHFFFAOYSA-N
SMILESCCCCCCC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5510-1.5550 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only