2,5-Dibromothiazole, 97%
2,5-Dibromothiazole, 97%
2,5-Dibromothiazole, 97%
Thermo Scientific Chemicals

2,5-Dibromothiazole, 97%

CAS: 4175-78-4 | C3HBr2NS | 242.916 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H26909.14
also known as H26909-14
Price (USD)
389.65
Online Exclusive
433.00
Save 43.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
389.65
Online Exclusive
433.00
Save 43.35 (10%)
Each
Chemical Identifiers
CAS4175-78-4
IUPAC Name2,5-dibromo-1,3-thiazole
Molecular FormulaC3HBr2NS
InChI KeyXIBIQFJKUZZLLX-UHFFFAOYSA-N
SMILESBrC1=CN=C(Br)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale brown
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)44.0-53.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only