1-Methyl-3-phenylpiperazine, 97%
1-Methyl-3-phenylpiperazine, 97%
1-Methyl-3-phenylpiperazine, 97%
Thermo Scientific Chemicals

1-Methyl-3-phenylpiperazine, 97%

CAS: 5271-27-2 | C11H16N2 | 176.263 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H26924.03
also known as H26924-03
Price (USD)
26.65
Online Exclusive
29.00
Save 2.35 (8%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
26.65
Online Exclusive
29.00
Save 2.35 (8%)
Each
Chemical Identifiers
CAS5271-27-2
IUPAC Name(2R)-4-methyl-2-phenylpiperazine-1,4-diium
Molecular FormulaC11H18N2
InChI KeyIRMBVBDXXYXPEW-NSHDSACASA-P
SMILESC[NH+]1CC[NH2+][C@@H](C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Melting Point (clear melt)54.0-63.0?C
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only