2-(3,4-Dimethoxyphenyl)ethanol, 98%
2-(3,4-Dimethoxyphenyl)ethanol, 98%
2-(3,4-Dimethoxyphenyl)ethanol, 98%
2-(3,4-Dimethoxyphenyl)ethanol, 98%
Thermo Scientific Chemicals

2-(3,4-Dimethoxyphenyl)ethanol, 98%

CAS: 7417-21-2 | C10H14O3 | 182.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number H26941.18
also known as H26941-18
Price (USD)
249.65
Special offer
Online exclusive
Ends: 30-Jun-2026
294.00
Save 44.35 (15%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
249.65
Special offer
Online exclusive
Ends: 30-Jun-2026
294.00
Save 44.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7417-21-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or pale cream
FormPowder or chunks
Melting Point (clear melt)44.0-50.0?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only