1-Allyl-3-methylimidazolium chloride, 98%
1-Allyl-3-methylimidazolium chloride, 98%
1-Allyl-3-methylimidazolium chloride, 98%
1-Allyl-3-methylimidazolium chloride, 98%
Thermo Scientific Chemicals

1-Allyl-3-methylimidazolium chloride, 98%

CAS: 65039-10-3 | C7H11ClN2 | 158.629 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number H26952.06
also known as H26952-06
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS65039-10-3
IUPAC Name3-methyl-1-(prop-2-en-1-yl)-1H-imidazol-3-ium chloride
Molecular FormulaC7H11ClN2
InChI KeyQVRCRKLLQYOIKY-UHFFFAOYSA-M
SMILES[Cl-].C[N+]1=CN(CC=C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)45-56?C
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Appearance (Color)White to cream to yellow to pale brown
FormCrystalline or fused solid or crystals or powder or lumps or crystalline powder
Identification (FTIR)Conforms
1-Allyl-3-methylimidazolium chloride is used as an ionic liquid as well as nonderivatizing solvent for cellulose. It acts as a plasticizer for cornstarch and finds application as a solid biopolymer electrolyte. Further, it plays an important role in the preparation of reducing sugar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Allyl-3-methylimidazolium chloride is used as an ionic liquid as well as nonderivatizing solvent for cellulose. It acts as a plasticizer for cornstarch and finds application as a solid biopolymer electrolyte. Further, it plays an important role in the preparation of reducing sugar.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Wang, J.; Zhou, M.; Yuan, Y.; Zhang, Q.; Fang, X.; Zang, S. Hydrolysis of cellulose catalyzed by quaternary ammonium perrhenates in 1-allyl-3-methylimidazolium chloride. Bioresour. Technol. 2015, 197, 42-47.
  2. Lv, F.; Wang, C.; Zhu, P.; Zhang, C. Isolation and recovery of cellulose from waste nylon/cotton blended fabrics by 1-allyl-3-methylimidazolium chloride. Carbohydr. Polym. 2015, 123, 424-431.