4-tert-Butylbenzyl bromide, 97%
4-tert-Butylbenzyl bromide, 97%
4-tert-Butylbenzyl bromide, 97%
4-tert-Butylbenzyl bromide, 97%
Thermo Scientific Chemicals

4-tert-Butylbenzyl bromide, 97%

CAS: 18880-00-7 | C11H15Br | 227.145 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H26991.06
also known as H26991-06
Price (USD)
34.65
Online Exclusive
38.00
Save 3.35 (9%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
34.65
Online Exclusive
38.00
Save 3.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS18880-00-7
IUPAC Name1-(bromomethyl)-4-tert-butylbenzene
Molecular FormulaC11H15Br
InChI KeyQZNQSIHCDAGZIA-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
Refractive Index1.5435-1.5485 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only