5,6-Dihydroxyflavone, 97%
5,6-Dihydroxyflavone, 97%
5,6-Dihydroxyflavone, 97%
5,6-Dihydroxyflavone, 97%
Thermo Scientific Chemicals

5,6-Dihydroxyflavone, 97%

CAS: 6665-66-3 | C15H10O4 | 254.241 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
5 g
Catalog number H27039.06
also known as H27039-06
Price (USD)
695.65
Special offer
Online exclusive
Ends: 30-Jun-2026
818.00
Save 122.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
695.65
Special offer
Online exclusive
Ends: 30-Jun-2026
818.00
Save 122.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS6665-66-3
IUPAC Name5,6-dihydroxy-2-phenyl-4H-chromen-4-one
Molecular FormulaC15H10O4
InChI KeyAGZAGADSYIYYCT-UHFFFAOYSA-N
SMILESOC1=CC=C2OC(=CC(=O)C2=C1O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Appearance (Color)Yellow
FormPowder
Melting Point (clear melt)185.5-191.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only