(S)-(+)-1-Bromo-2-methylbutane, 97%, stab. with potassium carbonate
(S)-(+)-1-Bromo-2-methylbutane, 97%, stab. with potassium carbonate
(S)-(+)-1-Bromo-2-methylbutane, 97%, stab. with potassium carbonate
Thermo Scientific Chemicals

(S)-(+)-1-Bromo-2-methylbutane, 97%, stab. with potassium carbonate

CAS: 534-00-9 | C5H11Br | 151.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H27078.06
also known as H27078-06
Price (USD)
153.65
Online Exclusive
170.00
Save 16.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
153.65
Online Exclusive
170.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS534-00-9
IUPAC Name(2S)-1-bromo-2-methylbutane
Molecular FormulaC5H11Br
InChI KeyXKVLZBNEPALHIO-YFKPBYRVSA-N
SMILESCC[C@H](C)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
CommentStabilised with 0.1% potassium carbonate, appears as sediment
Identification (FTIR)Conforms
View more
(S)-(+)-1-Bromo-2-methylbutane, is used for the synthesis of chiral nematic liquid crystals and optically active Grignard reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-1-Bromo-2-methylbutane, is used for the synthesis of chiral nematic liquid crystals and optically active Grignard reagents.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases. Light sensitive.
RUO – Research Use Only

General References:

  1. J Howarth.; K Hanlon.; D Fayne.; P McCormac. Moisture stable dialkylimidazolium salts as heterogeneous and homogeneous Lewis acids in the Diels-Alder reaction. Tetrahedron letters. 199738 (17), 3097-3100.
  2. PS Skell.; DL Tuleen.; PD Readio. The Mechanism of Aliphatic Bromination by N-Bromosuccinimide. 196385 (18), 2850-2851.