N-Boc-trans-4-hydroxy-L-proline, 97%
N-Boc-trans-4-hydroxy-L-proline, 97%
N-Boc-trans-4-hydroxy-L-proline, 97%
Thermo Scientific Chemicals

N-Boc-trans-4-hydroxy-L-proline, 97%

CAS: 13726-69-7 | C10H17NO5 | 231.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27110.06
also known as H27110-06
Price (USD)
86.65
Online exclusive
96.10
Save 9.45
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
86.65
Online exclusive
96.10
Save 9.45
Each
Chemical Identifiers
CAS13726-69-7
IUPAC Name1-[(tert-butoxy)carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid
Molecular FormulaC10H17NO5
InChI KeyBENKAPCDIOILGV-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CC(O)CC1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)>96.0%
Optical Rotation-53.5? ?2? (c=1 in DMF)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only