2-Nitroterephthalic acid 1-methyl ester, 97%
2-Nitroterephthalic acid 1-methyl ester, 97%
2-Nitroterephthalic acid 1-methyl ester, 97%
Thermo Scientific Chemicals

2-Nitroterephthalic acid 1-methyl ester, 97%

CAS: 35092-89-8 | C9H7NO6 | 225.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27121.06
also known as H27121-06
Price (USD)
23.80
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
23.80
Each
Chemical Identifiers
CAS35092-89-8
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Melting Point (clear melt)171.0-181.0?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only