2,6-Dimethoxyaniline, 97%
2,6-Dimethoxyaniline, 97%
2,6-Dimethoxyaniline, 97%
2,6-Dimethoxyaniline, 97%
Thermo Scientific Chemicals

2,6-Dimethoxyaniline, 97%

CAS: 2734-70-5 | C8H11NO2 | 153.181 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27139.06
also known as H27139-06
Price (USD)
268.65
Online Exclusive
298.00
Save 29.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
268.65
Online Exclusive
298.00
Save 29.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS2734-70-5
IUPAC Name2,6-dimethoxyaniline
Molecular FormulaC8H11NO2
InChI KeyHQBJSEKQNRSDAZ-UHFFFAOYSA-N
SMILESCOC1=CC=CC(OC)=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)72.0-82.0?C
2,6-Dimethylaniline degradation by Fenton process has been studied in depth for the purpose of learning more about the reactions involved in the oxidation of 2,6-dimethylaniline under various reaction conditions. Using a previously developed gas chromatographic-mass spectrometric assay, hemoglobin adducts of 2,6-dimethylaniline were detected covalently bound to rat hemoglobin after administration of either 2,6-dimethylaniline or lidocaine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylaniline degradation by Fenton process has been studied in depth for the purpose of learning more about the reactions involved in the oxidation of 2,6-dimethylaniline under various reaction conditions. Using a previously developed gas chromatographic-mass spectrometric assay, hemoglobin adducts of 2,6-dimethylaniline were detected covalently bound to rat hemoglobin after administration of either 2,6-dimethylaniline or lidocaine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Nalinrut Masomboon,; Chavalit Ratanatamskul,; Ming-Chun Lu. Chemical Oxidation of 2,6-Dimethylaniline in the Fenton Process. Environ. Sci. Technol.,. 2009, 43 (22) ,8629-8634.
  2. Matthew S. Bryant,; Henry F. Simmons,; Robert E. Harrell,; Jack A. Hinson. 2,6-Dimethylaniline—hemoglobin adducts from lidocaine in humans. Journal name. 1994, 15 (10),2287-2290..