1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride, 97%
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride, 97%
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride, 97%
Thermo Scientific Chemicals

1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride, 97%

CAS: 250285-32-6 | C27H37ClN2 | 425.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27150.03
also known as H27150-03
Price (USD)
165.65
Online Exclusive
184.00
Save 18.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
165.65
Online Exclusive
184.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS250285-32-6
IUPAC Name1,3-bis[2,6-bis(propan-2-yl)phenyl]-3H-1λ⁵-imidazol-1-ylium chloride
Molecular FormulaC27H37ClN2
InChI KeyAVJBQMXODCVJCJ-UHFFFAOYSA-M
SMILES[Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1C=C[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or pale pink
FormPowder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Water Content (Karl Fischer Titration)≤3%
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride is also used in organic synthesis, as well as a pharmaceutical intermediate. 1,3-Bis(2,6-diisopropylphenyl)imidazolium Chloride is used as a reagent in the synthesis of NHC Copper(I) complexes bearing dipyridylamine ligands which exhibit interesting luminescent properties and are potential candidates for organic light-emitting diode applications. 1,3-Bis(2,6-diisopropylphenyl)imidazolium Chloride is also used as a reagent in the synthesis of 5,6-Dimethyl-9-oxo-9H-xanthene-4-acetic Acid Methyl Ester (D476595); the methyl ester derivative of the drug Vadimezan (V084950).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride is also used in organic synthesis, as well as a pharmaceutical intermediate. 1,3-Bis(2,6-diisopropylphenyl)imidazolium Chloride is used as a reagent in the synthesis of NHC Copper(I) complexes bearing dipyridylamine ligands which exhibit interesting luminescent properties and are potential candidates for organic light-emitting diode applications. 1,3-Bis(2,6-diisopropylphenyl)imidazolium Chloride is also used as a reagent in the synthesis of 5,6-Dimethyl-9-oxo-9H-xanthene-4-acetic Acid Methyl Ester (D476595); the methyl ester derivative of the drug Vadimezan (V084950).

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only

General References:

  1. L Jafarpour.; ED Stevens.; SP Nolan. A sterically demanding nucleophilic carbene: 1, 3-bis (2, 6-diisopropylphenyl) imidazol-2-ylidene). Thermochemistry and catalytic application in olefin metathesis. Journal of Organometallic Chemistry. 2000606 (1), 49-54.
  2. J Huang.; SP Nolan. Efficient cross-coupling of aryl chlorides with aryl Grignard reagents (Kumada reaction) mediated by a palladium/imidazolium chloride system. J. Am. Chem. Soc. 2000121 (42), 9889-9890.
  3. In the presence of a base such as KO-t-Bu, generates the imidazol-2-ylidene, an example of an N-heterocyclic carbene (NHC). NHCs are relatively strong bases, and valuable replacements for phosphine ligands in transition metal chemistry. They are relatively stable to dimerization, but sensitive to air and moisture, so are best generated in situ. Review: Angew. Chem. Int. Ed., 36, 2163 (1997). For a review of stable carbenes, see: Chem. Rev., 100, 39 (2000).
  4. Used as a phosphine-free ligand in various metal-catalyzed coupling reactions, often with advantageous results in difficult cases. For use in the Pd-catalyzed cross-coupling of aryl Grignards with aryl chlorides (Kumada reaction), see: J. Am. Chem. Soc., 121, 9889 (1999). For Pd-catalyzed coupling of trimethoxysilanes with electron-deficient aryl chlorides, see: Org. Lett., 2, 2053 (2000). Many examples have been recorded of the use of NHC ligands in the Suzuki coupling reaction, for examples utilizing 1,3-bis(2,6-diisopropylphenyl)imiazol-2-ylidene, enabling otherwise difficult coupling reactions of aryl chlorides with aryl and alkyl boronic acids, see: Synlett, 292 (2001) (includes alkylboronic acids: better yields than 1,3-dimesitylimidazol-2-ylidene); Org. Lett., 6, 4435 (2004); Tetrahedron Lett., 45, 3511 (2004).
  5. See also 1,3-Dimesityl imidazolium chloride, H27535.