Methyldiphenylphosphine, 99%
Methyldiphenylphosphine, 99%
Methyldiphenylphosphine, 99%
Thermo Scientific Chemicals

Methyldiphenylphosphine, 99%

CAS: 1486-28-8 | C13H13P | 200.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Catalog number H27159.09
also known as H27159-09
Price (USD)
107.65
Special offer
Online exclusive
Ends: 30-Jun-2026
126.00
Save 18.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
107.65
Special offer
Online exclusive
Ends: 30-Jun-2026
126.00
Save 18.35 (15%)
Each
Chemical Identifiers
CAS1486-28-8
IUPAC Namemethyldiphenylphosphane
Molecular FormulaC13H13P
InChI KeyUJNZOIKQAUQOCN-UHFFFAOYSA-N
SMILESCP(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.6225-1.6275 @ 20°C
Appearance (Color)Clear colorless
Methyldiphenylphosphine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyldiphenylphosphine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only

General References:

  1. DA Redfield.; LW Cary.; JH Nelson. Equilibrium thermodynamics and mechanism of cis-trans isomerization for diazidobis(methyldiphenylphosphine)palladium(II) and diazidobis(dimethylphenylphosphine)palladium(II). Inorganic Chemistry. 197514 (1), 50-59.
  2. DA Redfield.; LW Cary.; JH Nelson. Equilibrium thermodynamics and mechanism of cis-trans isomerization for diazidobis(methyldiphenylphosphine)palladium(II) and diazidobis(dimethylphenylphosphine)palladium(II). J. Chem. Soc., Chem. Commun. 198114 (1), 50-59.