6-Nitrochromone, 97%
6-Nitrochromone, 97%
6-Nitrochromone, 97%
Thermo Scientific Chemicals

6-Nitrochromone, 97%

CAS: 51484-05-0 | C9H5NO4 | 191.142 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H27167.03
also known as H27167-03
Price (USD)
23.65
Online exclusive
25.90
Save 2.25
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
23.65
Online exclusive
25.90
Save 2.25
Each
Chemical Identifiers
CAS51484-05-0
IUPAC Name6-nitro-4H-chromen-4-one
Molecular FormulaC9H5NO4
InChI KeyORWADBVBOPTYQT-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C2OC=CC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)169.5-178.5?C
Appearance (Color)White to yellow or pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only