1-Ethyl-3-methylimidazolium hydrogen sulfate, 98%
1-Ethyl-3-methylimidazolium hydrogen sulfate, 98%
1-Ethyl-3-methylimidazolium hydrogen sulfate, 98%
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium hydrogen sulfate, 98%

CAS: 412009-61-1 | C6H12N2O4S | 208.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number H27232.30
also known as H27232-30
Price (USD)
379.65
Special offer
Online exclusive
Ends: 30-Jun-2026
446.00
Save 66.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
379.65
Special offer
Online exclusive
Ends: 30-Jun-2026
446.00
Save 66.35 (15%)
Each
Chemical Identifiers
CAS412009-61-1
IUPAC Name1-ethyl-3-methyl-1H-imidazol-3-ium hydrogen sulfate
Molecular FormulaC6H12N2O4S
InChI KeyHZKDSQCZNUUQIF-UHFFFAOYSA-M
SMILESOS([O-])(=O)=O.CCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)> 97.5% by ion chromatography
Water Content (Karl Fischer Titration)< 0.5%
FormLiquid
Proton NMRTo conform to structure
Appearance (Color)Clear colourless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only