2-Mercapto-5-n-propylpyrimidine, 98%
2-Mercapto-5-n-propylpyrimidine, 98%
2-Mercapto-5-n-propylpyrimidine, 98%
2-Mercapto-5-n-propylpyrimidine, 98%
Thermo Scientific Chemicals

2-Mercapto-5-n-propylpyrimidine, 98%

CAS: 52767-84-7 | C7H10N2S | 154.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H27239.03
also known as H27239-03
Price (USD)
47.65
Online Exclusive
53.30
Save 5.65 (11%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
47.65
Online Exclusive
53.30
Save 5.65 (11%)
Each
Add to cart
Chemical Identifiers
CAS52767-84-7
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Appearance (Color)Yellow
Melting Point (clear melt)205.0-212.0?C
By using 2-mercapto-5-n-propylpyrimidine (MPP) as capping ligands, copper nanoclusters with different core sizes were prepared using a chemical reduction method. 2-Mercapto-5-n-propylpyrimidine finds use as means of synthesizing enantiomerically enriched compounds. Used as an odorant to help detect natural gas leaks. As intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
By using 2-mercapto-5-n-propylpyrimidine (MPP) as capping ligands, copper nanoclusters with different core sizes were prepared using a chemical reduction method. 2-Mercapto-5-n-propylpyrimidine finds use as means of synthesizing enantiomerically enriched compounds. Used as an odorant to help detect natural gas leaks. As intermediate in organic syntheses.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Wentao Weia,; Wei Chena. Size-dependent catalytic activity of copper nanoclusters for oxygen electroreduction in alkaline solution . International Journal of Smart and Nano Materials. 2013, 4 (1), 62-71.
  2. Shradha. S. Binani,; P. S. bodke,; R.V.Joat. Synthesis, Characterization and In vitro Antimicrobial Evaluation of Novel 2- Mercapto-4,6-Disubstituted Phenyl Pyrimidine Derivatives. International Journal of Pharmacy and Pharmaceutical Sciences. 2014 , 6 (1), 1, 461-463.