1-n-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
1-n-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
1-n-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
Thermo Scientific Chemicals

1-n-Hexyl-3-methylimidazolium hexafluorophosphate, 99%

CAS: 304680-35-1 | C10H19F6N2P | 312.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number H27272.06
also known as H27272-06
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS304680-35-1
IUPAC Name1-hexyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC10H19F6N2P
InChI KeyYPWSSSRXUOQNMQ-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4165-1.4200 @ 20?C
Appearance (Color)Clear, colorless to pale yellow or yellow to orange
Residual halide<250ppm
Water Content (Karl Fischer Titration)<250ppm
FormLiquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only