2,6-Diisopropyl-N,N-dimethylaniline, 96%
2,6-Diisopropyl-N,N-dimethylaniline, 96%
2,6-Diisopropyl-N,N-dimethylaniline, 96%
Thermo Scientific Chemicals

2,6-Diisopropyl-N,N-dimethylaniline, 96%

CAS: 2909-77-5 | C14H23N | 205.345 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27275.06
also known as H27275-06
Price (USD)
90.65
Online Exclusive
101.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
90.65
Online Exclusive
101.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS2909-77-5
IUPAC NameN,N-dimethyl-2,6-bis(propan-2-yl)aniline
Molecular FormulaC14H23N
InChI KeyALXIOUGHHXXLKX-UHFFFAOYSA-N
SMILESCC(C)C1=CC=CC(C(C)C)=C1N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥95.0%
Refractive Index1.4970-1.5030 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only