4-Bromo-2-chloropyridine, 94%
4-Bromo-2-chloropyridine, 94%
4-Bromo-2-chloropyridine, 94%
Thermo Scientific Chemicals

4-Bromo-2-chloropyridine, 94%

CAS: 73583-37-6 | C5H3BrClN | 192.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27299.06
also known as H27299-06
Price (USD)
32.65
Online Exclusive
36.30
Save 3.65 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
32.65
Online Exclusive
36.30
Save 3.65 (10%)
Each
Chemical Identifiers
CAS73583-37-6
IUPAC Name4-bromo-2-chloropyridine
Molecular FormulaC5H3BrClN
InChI KeyONHMWUXYIFULDO-UHFFFAOYSA-N
SMILESClC1=CC(Br)=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥92.0%
Refractive Index1.5835-1.5905 @ 20?C
It is used in the synthesis of quaterpyridine nemertelline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of quaterpyridine nemertelline.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Andreas Reichelt, et. al. Palladium-Catalyzed Chemoselective Monoarylation of Hydrazides for the Synthesis of [1,2,4]Triazolo[4,3-a]pyridines.Org. Lett.,2003,68(26), 10178-10180.
  2. Alexandre Bouillon, et al. An Efficient Two-Step Total Synthesis of the Quaterpyridine Nemertelline.J. Org. Chem.,2008,49(33), 4862-4864.