2-Bromo-6-nitrophenol, 97%
2-Bromo-6-nitrophenol, 97%
2-Bromo-6-nitrophenol, 97%
Thermo Scientific Chemicals

2-Bromo-6-nitrophenol, 97%

CAS: 13073-25-1 | C6H4BrNO3 | 218.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27326.14
also known as H27326-14
Price (USD)
306.65
Online Exclusive
341.00
Save 34.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
306.65
Online Exclusive
341.00
Save 34.35 (10%)
Each
Chemical Identifiers
CAS13073-25-1
IUPAC Name2-bromo-6-nitrophenol
Molecular FormulaC6H4BrNO3
InChI KeyVEJSIOPQKQXJAT-UHFFFAOYSA-N
SMILESOC1=C(Br)C=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown
Melting Point (clear melt)63.0-72.0?C
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only