1-n-Butyl-4-methylpyridinium hexafluorophosphate, 99%
1-n-Butyl-4-methylpyridinium hexafluorophosphate, 99%
1-n-Butyl-4-methylpyridinium hexafluorophosphate, 99%
Thermo Scientific Chemicals

1-n-Butyl-4-methylpyridinium hexafluorophosphate, 99%

CAS: 401788-99-6 | C10H16F6NP | 295.209 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number H27355.18
also known as H27355-18
Price (USD)
266.65
Special offer
Online exclusive
Ends: 30-Jun-2026
314.00
Save 47.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
266.65
Special offer
Online exclusive
Ends: 30-Jun-2026
314.00
Save 47.35 (15%)
Each
Chemical Identifiers
CAS401788-99-6
IUPAC Name1-butyl-4-methylpyridin-1-ium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC10H16F6NP
InChI KeyXBSZSSGZQFAUPF-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CCCC[N+]1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Water Content (Karl Fischer Titration)≤1,000ppm (≤0.1000%)
CommentHexafluorophosphate (anion) content: ≥98.5% (ion chromatography)
Residual halide≤100ppm (ion chromatography)
Appearance (Color)Yellow to brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only