2-Chloro-D-phenylalanine, 98%
2-Chloro-D-phenylalanine, 98%
2-Chloro-D-phenylalanine, 98%
2-Chloro-D-phenylalanine, 98%
Thermo Scientific Chemicals

2-Chloro-D-phenylalanine, 98%

CAS: 80126-50-7 | C9H10ClNO2 | 199.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27359.03
also known as H27359-03
Price (USD)
138.65
Online Exclusive
154.00
Save 15.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
138.65
Online Exclusive
154.00
Save 15.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS80126-50-7
IUPAC Name2-amino-3-(2-chlorophenyl)propanoic acid
Molecular FormulaC9H10ClNO2
InChI KeyCVZZNRXMDCOHBG-UHFFFAOYNA-N
SMILESNC(CC1=CC=CC=C1Cl)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)209.0-215.0?C
Enantiomeric excess≥97.5%
Assay (HPLC)≥97.5%
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only