4-Aminopyrimidine, 98%
4-Aminopyrimidine, 98%
4-Aminopyrimidine, 98%
Thermo Scientific Chemicals

4-Aminopyrimidine, 98%

CAS: 591-54-8 | C4H5N3 | 95.105 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H27367.03
also known as H27367-03
Price (USD)
125.65
Online Exclusive
139.00
Save 13.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
125.65
Online Exclusive
139.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS591-54-8
IUPAC Namepyrimidin-4-amine
Molecular FormulaC4H5N3
InChI KeyOYRRZWATULMEPF-UHFFFAOYSA-N
SMILESNC1=CC=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to cream to yellow to pale brown
Melting Point (clear melt)146.0-153.0?C
FormCrystals or powder or crystalline powder
4-Aminopyrimidine has been used in the preparation of 1:4-dihydro-4-imino-1-methylpyrimidine hydriodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminopyrimidine has been used in the preparation of 1:4-dihydro-4-imino-1-methylpyrimidine hydriodide.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Johan Smets, et al. Matrix-Isolation FT-IR Studies and Ab-Initio Calculations of Hydrogen-Bonded Complexes of Molecules Modeling Cytosine or Isocytosine Tautomers. 2. 4-Aminopyridine and 4-Aminopyrimidine Complexes with H2O in Ar Matrixes.J. Phys. Chem.,1995,99(17), 6387-6400.
  2. Eixjardo Scarano, et al. The Enzymatic Aminohydrolysis of 4-Aminopyrimidine Deoxyribonucleotides.J. Biol. Chem.,1963,238PC1556-PC1557.