N-Boc-4-oxo-L-proline tert-butyl ester, 97%
N-Boc-4-oxo-L-proline tert-butyl ester, 97%
N-Boc-4-oxo-L-proline tert-butyl ester, 97%
Thermo Scientific Chemicals

N-Boc-4-oxo-L-proline tert-butyl ester, 97%

CAS: 166410-05-5 | C14H23NO5 | 285.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27375.06
also known as H27375-06
Price (USD)
595.65
Special offer
Online exclusive
Ends: 30-Jun-2026
700.00
Save 104.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
595.65
Special offer
Online exclusive
Ends: 30-Jun-2026
700.00
Save 104.35 (15%)
Each
Chemical Identifiers
CAS166410-05-5
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Optical Rotation+9? ± 1.0? (c=1 in chloroform)
Appearance (Color)Pale cream
Melting Point (clear melt)61.0-70.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only