Potassium 4-methoxyphenyltrifluoroborate, 95%
Potassium 4-methoxyphenyltrifluoroborate, 95%
Potassium 4-methoxyphenyltrifluoroborate, 95%
Thermo Scientific Chemicals

Potassium 4-methoxyphenyltrifluoroborate, 95%

CAS: 192863-36-8 | C7H7BF3KO | 214.036 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27384.06
also known as H27384-06
Price (USD)
159.65
Special offer
Online exclusive
Ends: 30-Jun-2026
188.00
Save 28.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
159.65
Special offer
Online exclusive
Ends: 30-Jun-2026
188.00
Save 28.35 (15%)
Each
Chemical Identifiers
CAS192863-36-8
IUPAC Namepotassium trifluoro(4-methoxyphenyl)boranuide
Molecular FormulaC7H7BF3KO
InChI KeyXNYMCUFKZHRYID-UHFFFAOYSA-N
SMILES[K+].COC1=CC=C(C=C1)[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (HPLC)≥94.0%
It is applied in Suzuki Cross-Coupling using organotrifluoroborates as potent boronic acid surrogates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in Suzuki Cross-Coupling using organotrifluoroborates as potent boronic acid surrogates.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. George W. Kabalka, et al. A facile synthesis of aryl iodides via potassium aryltrifluoroborates.Tetrahedron Lett.,2004,45(2), 343-345.
  2. Rafael C, et al. Chemoselective cross-coupling Suzuki-Miyaura reaction of (Z)-(2-chlorovinyl)tellurides and potassium aryltrifluoroborate salts.Tetrahedron Lett.,2008,49(32), 4713-4716.