4-Benzyloxy-6-chloropyrimidine, 95%
4-Benzyloxy-6-chloropyrimidine, 95%
4-Benzyloxy-6-chloropyrimidine, 95%
4-Benzyloxy-6-chloropyrimidine, 95%
Thermo Scientific Chemicals

4-Benzyloxy-6-chloropyrimidine, 95%

CAS: 405930-65-6 | C11H9ClN2O | 220.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27390.03
also known as H27390-03
Price (USD)
298.65
Online Exclusive
332.00
Save 33.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
298.65
Online Exclusive
332.00
Save 33.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS405930-65-6
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
FormPowder
Melting Point (clear melt)29.5-38.5?C
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only