1-Ethyl-3-methylimidazolium diethyl phosphate, 98%
1-Ethyl-3-methylimidazolium diethyl phosphate, 98%
1-Ethyl-3-methylimidazolium diethyl phosphate, 98%
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium diethyl phosphate, 98%

CAS: 848641-69-0 | C10H21N2O4P | 264.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27400.06
also known as H27400-06
Price (USD)
49.65
Online Exclusive
54.90
Save 5.25 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
49.65
Online Exclusive
54.90
Save 5.25 (10%)
Each
Chemical Identifiers
CAS848641-69-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow or red
Water Content (Karl Fischer Titration)<1.0%
Assay (HPLC)≥97.5%
Refractive Index1.4705-1.4755 @ 20?C
FormLiquid
It finds its application in improving enzymatic hydrolysis of wheat straw and in the enzymatic in situ saccharification of cellulose in aqueous-ionic liquid media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in improving enzymatic hydrolysis of wheat straw and in the enzymatic in situ saccharification of cellulose in aqueous-ionic liquid media.

Solubility
Fully miscible in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. Paetrice O. Jones, et al. Cellulose hydrolysis by immobilized Trichoderma reesei cellulase.Biotechnol. Lett.,2010,32(1), 103-106.
  2. Jürgen Vitz, et al. Extended dissolution studies of cellulose in imidazolium based ionic liquids.Green Chem.,2009,11(3), 417-424.