2,6-Dichloropyridine-3-boronic acid, 95%
2,6-Dichloropyridine-3-boronic acid, 95%
2,6-Dichloropyridine-3-boronic acid, 95%
Thermo Scientific Chemicals

2,6-Dichloropyridine-3-boronic acid, 95%

CAS: 148493-34-9 | C5H4BCl2NO2 | 191.802 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27406.03
also known as H27406-03
Price (USD)
209.65
Online Exclusive
233.00
Save 23.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
209.65
Online Exclusive
233.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS148493-34-9
IUPAC Name(2,6-dichloropyridin-3-yl)boronic acid
Molecular FormulaC5H4BCl2NO2
InChI KeyXBBLBQZAVMHEER-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(Cl)N=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥94.0%
Proton NMRConforms to structure
FormPowder
Melting Point (clear melt)91.0-103.0?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only