6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, 95%
6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, 95%
6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, 95%
Thermo Scientific Chemicals

6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, 95%

CAS: 1032758-87-4 | C17H27BN2O4 | 334.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27430.MD
also known as H27430-MD
Price (USD)
101.00
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
101.00
Each
Chemical Identifiers
CAS1032758-87-4
IUPAC Nametert-butyl N-methyl-N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
Molecular FormulaC17H27BN2O4
InChI KeyQGBJKHALDLKAGX-UHFFFAOYSA-N
SMILESCN(C(=O)OC(C)(C)C)C1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay by NMR>94.0%
Proton NMRConforms to structure
Used in the synthesis of organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of organic compounds.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Michael Königa,; Lorenz Michael Reitha,; Uwe Monkowiusa,; Günther Knöra,; Klaus Bretterbauerb,; Wolfgang Schoefberger. 6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, 95%. Tetrahedron. 2011 , 67(23),4243-4252.