5-Amino-2-methylpyridine, 97%
5-Amino-2-methylpyridine, 97%
5-Amino-2-methylpyridine, 97%
Thermo Scientific Chemicals

5-Amino-2-methylpyridine, 97%

CAS: 3430-14-6 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27431.MD
also known as H27431-MD
Price (USD)
74.65
Online exclusive
82.30
Save 7.65
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
74.65
Online exclusive
82.30
Save 7.65
Each
Chemical Identifiers
CAS3430-14-6
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤2.25%
Assay (GC)≥96.0%
Melting Point (clear melt)85.0-97.0?C
Appearance (Color)Brown
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only