6-Bromo-2,4-dimethylpyridine-3-boronic acid, 95%
6-Bromo-2,4-dimethylpyridine-3-boronic acid, 95%
6-Bromo-2,4-dimethylpyridine-3-boronic acid, 95%
6-Bromo-2,4-dimethylpyridine-3-boronic acid, 95%
Thermo Scientific Chemicals

6-Bromo-2,4-dimethylpyridine-3-boronic acid, 95%

CAS: 1072944-23-0 | C7H9BBrNO2 | 229.868 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27460.03
also known as H27460-03
Price (USD)
467.65
Special offer
Online exclusive
Ends: 30-Jun-2026
550.00
Save 82.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
467.65
Special offer
Online exclusive
Ends: 30-Jun-2026
550.00
Save 82.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS1072944-23-0
IUPAC Name(6-bromo-2,4-dimethylpyridin-3-yl)boronic acid
Molecular FormulaC7H9BBrNO2
InChI KeyGXXITYQLWOMIEM-UHFFFAOYSA-N
SMILESCC1=CC(Br)=NC(C)=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥94.0%
Appearance (Color)White to pale cream
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only